2-(tert-Butyl)isonicotinic acid 95%

Lieferant: Apollo Scientific

OR943509-1G OR943509-250MG
APOSOR943509-1GEA 212 EUR
APOSOR943509-1G APOSOR943509-250MG
2-(tert-Butyl)isonicotinic acid 95%
2-(tert-Butyl)isonicotinic acid
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04113815
CAS-Nummer: 91940-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR