6-Cyano-4-chromanone 95%

Lieferant: Apollo Scientific

OR943666-1G OR943666-250MG
APOSOR943666-1GEA 257 EUR
APOSOR943666-1G APOSOR943666-250MG
6-Cyano-4-chromanone 95%
6-Cyano-4-chromanone
Formel: C₁₀H₇NO₂
Molecular Weight: 173,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05861625
CAS-Nummer: 214759-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR