Methyl 2-[2-(chloromethyl)phenyl]acetate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(2-(chloromethyl)phenyl)acetate

OR943804-1G OR943804-250MG
APOSOR943804-1GEA 647 EUR
APOSOR943804-1G APOSOR943804-250MG
Methyl 2-[2-(chloromethyl)phenyl]acetate 95%
Methyl 2-[2-(chloromethyl)phenyl]acetate
Formel: C₁₀H₁₁ClO₂
Molecular Weight: 198,646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832998
CAS-Nummer: 95360-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR