3-Ethynylpyridin-2-amine 95%
Lieferant: Apollo Scientific
OR944028-1G
OR944028-250MG
APOSOR944028-1GEA
440
EUR
APOSOR944028-1G
APOSOR944028-250MG
3-Ethynylpyridin-2-amine 95%
3-Ethynylpyridin-2-amine
Formel:
C₇H₆N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13175219 CAS-Nummer: 67346-74-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...