4-Hydroxy-2,3-dimethylbenzonitrile 95%

Lieferant: Apollo Scientific

OR944047-1G OR944047-250MG
APOSOR944047-1GEA 366 EUR
APOSOR944047-1G APOSOR944047-250MG
4-Hydroxy-2,3-dimethylbenzonitrile 95%
4-Hydroxy-2,3-dimethylbenzonitrile
Formel: C₉H₉NO
Molecular Weight: 147,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13659357
CAS-Nummer: 448961-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR