N-Methyl 4-chloro-2-nitroaniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-N-methyl-2-nitroaniline

OR9445-1G OR9445-5G
APOSOR9445-1GEA 39.5 EUR
APOSOR9445-1G APOSOR9445-5G
N-Methyl 4-chloro-2-nitroaniline
N-Methyl 4-chloro-2-nitroaniline
Formel: C₇H₇ClN₂O₂
Molecular Weight: 186.596 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00461746
CAS-Nummer: 15950-17-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR