2-(4-Methylcyclohexyl)acetic acid 95%
Lieferant: Apollo Scientific
OR944568-1G
OR944568-250MG
APOSOR944568-1GEA
128
EUR
APOSOR944568-1G
APOSOR944568-250MG
2-(4-Methylcyclohexyl)acetic acid 95%
2-(4-Methylcyclohexyl)acetic acid
Formel:
C₉H₁₆O₂ Molecular Weight: 156,222 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074993 CAS-Nummer: 6603-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...