2-Phenylcyclopropylamine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Phenylcyclopropan-1-amine

OR944624-1G OR944624-5G
APOSOR944624-1GEA 190 EUR
APOSOR944624-1G APOSOR944624-5G
2-Phenylcyclopropylamine 97%
2-Phenylcyclopropylamine
Formel: C₉H₁₁N
Lagertemperatur: Kühlschrank
MDL: MFCD04114536
CAS-Nummer: 54-97-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR