tert-Butyl piperidine-4-carboxylate 95%

Lieferant: Apollo Scientific

OR944680-1G OR944680-250MG
APOSOR944680-1GEA 156 EUR
APOSOR944680-1G APOSOR944680-250MG
tert-Butyl piperidine-4-carboxylate 95%
tert-Butyl piperidine-4-carboxylate
Formel: C₁₀H₁₉NO₂
Molecular Weight: 185.263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684429
CAS-Nummer: 138007-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR