6-Bromo-2-(methylsulphonyl)-1,2,3,4-tetrahydroisoquinoline 95%

Lieferant: Apollo Scientific

OR945026-1G OR945026-250MG
APOSOR945026-1GEA 267 EUR
APOSOR945026-1G APOSOR945026-250MG
6-Bromo-2-(methylsulphonyl)-1,2,3,4-tetrahydroisoquinoline 95%
6-Bromo-2-(methylsulphonyl)-1,2,3,4-tetrahydroisoquinoline
Formel: C₁₀H₁₂BrNO₂S
Molecular Weight: 290,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109847
CAS-Nummer: 1046816-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR