(R)-1-Boc-2-[methoxy(methyl)carbamoyl]pyrrolidine 97%
Lieferant: Apollo Scientific
OR945542-1G
OR945542-500MG
OR945542-5G
APOSOR945542-1GEA
261
EUR
APOSOR945542-1G
APOSOR945542-500MG
APOSOR945542-5G
(R)-1-Boc-2-[methoxy(methyl)carbamoyl]pyrrolidine 97%
(R)-1-Boc-2-[methoxy(methyl)carbamoyl]pyrrolidine
Formel:
C₁₂H₂₂N₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09756517 CAS-Nummer: 288086-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...