2-Chloro-6-methoxypyridin-4-amine 95+%

Lieferant: Apollo Scientific
Achtung

OR945855-250MG OR945855-1G
APOSOR945855-250MGEA 85.7 EUR
APOSOR945855-250MG APOSOR945855-1G
2-Chloro-6-methoxypyridin-4-amine 95+%
2-Chloro-6-methoxypyridin-4-amine
Formel: C₆H₇ClN₂O
Molecular Weight: 158.586 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13189324
CAS-Nummer: 1008304-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR