tert-Butyl 2-(methylsulphonyl)acetate 95%

Lieferant: Apollo Scientific

OR945903-1G OR945903-250MG
APOSOR945903-1GEA 435 EUR
APOSOR945903-1G APOSOR945903-250MG
tert-Butyl 2-(methylsulphonyl)acetate 95%
tert-Butyl 2-(methylsulphonyl)acetate
Formel: C₇H₁₄O₄S
Molecular Weight: 194,249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19632339
CAS-Nummer: 70018-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR