trans-3-(Benzyloxy)cyclobutanamine
Lieferant: Apollo Scientific
OR945984-1G
OR945984-250MG
OR945984-500MG
APOSOR945984-1GEA
899
EUR
APOSOR945984-1G
APOSOR945984-250MG
APOSOR945984-500MG
trans-3-(Benzyloxy)cyclobutanamine
trans-3-(Benzyloxy)cyclobutanamine
Formel:
C₁₁H₁₅NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD16660373 CAS-Nummer: 905821-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...