8-Chloroquinazolin-4(3H)-one 95%
Lieferant: Apollo Scientific
Synonyme:
8-Chloroquinazolin-4-ol
OR946042-1G
OR946042-5G
APOSOR946042-1GEA
66.7
EUR
APOSOR946042-1G
APOSOR946042-5G
8-Chloroquinazolin-4(3H)-one 95%
8-Chloroquinazolin-4(3H)-one
Formel:
C₈H₅ClN₂O Molecular Weight: 180,591 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09954861 CAS-Nummer: 101494-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...