8-Methoxyquinoline-2-carbaldehyde 95%

Lieferant: Apollo Scientific

OR946222-1G OR946222-250MG
APOSOR946222-1GEA 78.3 EUR
APOSOR946222-1G APOSOR946222-250MG
8-Methoxyquinoline-2-carbaldehyde 95%
8-Methoxyquinoline-2-carbaldehyde
Formel: C₁₁H₉NO₂
Molecular Weight: 187,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864557
CAS-Nummer: 103854-64-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR