6-Chloro-1,2-benzoxazol-3-amine 98%

Lieferant: Apollo Scientific

Synonyme: 6-Chlorobenzo[d]isoxazol-3-amine

OR946354-1G OR946354-5G
APOSOR946354-1GEA 458 EUR
APOSOR946354-1G APOSOR946354-5G
6-Chloro-1,2-benzoxazol-3-amine 98%
6-Chloro-1,2-benzoxazol-3-amine
Formel: C₇H₅ClN₂O
Molecular Weight: 168,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09055056
CAS-Nummer: 89692-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR