3-(Chloromethyl)-N-methylbenzamide 98%

Lieferant: Apollo Scientific

OR946371-10G OR946371-1G OR946371-5G
APOSOR946371-10GEA 451 EUR
APOSOR946371-10G APOSOR946371-1G APOSOR946371-5G
3-(Chloromethyl)-N-methylbenzamide 98%
3-(Chloromethyl)-N-methylbenzamide
Formel: C₉H₁₀ClNO
Molecular Weight: 183,635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11208364
CAS-Nummer: 123944-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR