4-(3-Methoxypropoxy)benzonitrile 98%
Lieferant: Apollo Scientific
OR946389-10G
OR946389-1G
OR946389-5G
APOSOR946389-10GEA
864
EUR
APOSOR946389-10G
APOSOR946389-1G
APOSOR946389-5G
4-(3-Methoxypropoxy)benzonitrile 98%
4-(3-Methoxypropoxy)benzonitrile
Formel:
C₁₁H₁₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11531994 CAS-Nummer: 943150-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...