N-Ethyl-2-(3-methoxyphenyl)acetamide 96%
Lieferant: Apollo Scientific
OR946457-1G
OR946457-5G
APOSOR946457-1GEA
203
EUR
APOSOR946457-1G
APOSOR946457-5G
N-Ethyl-2-(3-methoxyphenyl)acetamide 96%
N-Ethyl-2-(3-methoxyphenyl)acetamide
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193,242 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19595636 CAS-Nummer: 1342495-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...