(3-P-Tolyl-isoxazol-5-yl)-methanol 95%

Lieferant: Apollo Scientific

OR946560-1G OR946560-250MG
APOSOR946560-1GEA 841 EUR
APOSOR946560-1G APOSOR946560-250MG
(3-P-Tolyl-isoxazol-5-yl)-methanol 95%
(3-P-Tolyl-isoxazol-5-yl)-methanol
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738486
CAS-Nummer: 206055-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR