2-(5-Chloropyridin-2-yl)acetic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR946636-100MG OR946636-250MG
APOSOR946636-100MGEA 37.7 EUR
APOSOR946636-100MG APOSOR946636-250MG
2-(5-Chloropyridin-2-yl)acetic acid 95+%
2-(5-Chloropyridin-2-yl)acetic acid
Formel: C₇H₆ClNO₂
Molecular Weight: 171,581 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09925072
CAS-Nummer: 1000522-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR