5-Methoxynaphthalen-1-amine 95%

Lieferant: Apollo Scientific

OR947026-1G OR947026-250MG
APOSOR947026-1GEA 696 EUR
APOSOR947026-1G APOSOR947026-250MG
5-Methoxynaphthalen-1-amine 95%
5-Methoxynaphthalen-1-amine
Formel: C₁₁H₁₁NO
Molecular Weight: 173.211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00839435
CAS-Nummer: 6271-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR