1,3-Dimethoxy-4-methyl-2-nitrobenzene 95%

Lieferant: Apollo Scientific

OR947189-1G OR947189-250MG
APOSOR947189-1GEA 696 EUR
APOSOR947189-1G APOSOR947189-250MG
1,3-Dimethoxy-4-methyl-2-nitrobenzene 95%
1,3-Dimethoxy-4-methyl-2-nitrobenzene
Formel: C₉H₁₁NO₄
Molecular Weight: 197,188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14583015
CAS-Nummer: 1378831-08-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR