Dipiperidinomethane 95%

Lieferant: Apollo Scientific

OR947194-1G OR947194-5G
APOSOR947194-1GEA 1280 EUR
APOSOR947194-1G APOSOR947194-5G
Dipiperidinomethane 95%
Dipiperidinomethane
Formel: C₁₁H₂₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00006502
CAS-Nummer: 880-09-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR