2-(Formylamino)pyridine 95%
Lieferant: Apollo Scientific
OR947271-1G
OR947271-250MG
APOSOR947271-1GEA
841
EUR
APOSOR947271-1G
APOSOR947271-250MG
2-(Formylamino)pyridine 95%
2-(Formylamino)pyridine
Formel:
C₆H₆N₂O Molecular Weight: 122.125 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043008 CAS-Nummer: 34813-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...