2-(2,5-Dimethylphenyl)ethanol 95%
Lieferant: Apollo Scientific
OR947310-1G
OR947310-5G
APOSOR947310-1GEA
26.1
EUR
APOSOR947310-1G
APOSOR947310-5G
2-(2,5-Dimethylphenyl)ethanol 95%
2-(2,5-Dimethylphenyl)ethanol
Formel:
C₁₀H₁₄O Molecular Weight: 150,218 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020610 CAS-Nummer: 6972-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...