2-(2-Cyano-4-methoxyphenyl)acetic acid 95%

Lieferant: Apollo Scientific

OR947312-1G OR947312-250MG
APOSOR947312-1GEA 446 EUR
APOSOR947312-1G APOSOR947312-250MG
2-(2-Cyano-4-methoxyphenyl)acetic acid 95%
2-(2-Cyano-4-methoxyphenyl)acetic acid
Formel: C₁₀H₉NO₃
Molecular Weight: 191,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18394271
CAS-Nummer: 52786-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR