Methyl 5-formyl-2-methylbenzoate 95%

Lieferant: Apollo Scientific
Achtung

OR947357-1G OR947357-250MG
APOSOR947357-1GEA 509 EUR
APOSOR947357-1G APOSOR947357-250MG
Methyl 5-formyl-2-methylbenzoate 95%
Methyl 5-formyl-2-methylbenzoate
Formel: C₁₀H₁₀O₃
Molecular Weight: 178,185 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21099506
CAS-Nummer: 675148-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR