5-Chloro-2-formylbenzonitrile 95%

Lieferant: Apollo Scientific

OR947365-1G OR947365-250MG
APOSOR947365-1GEA 101 EUR
APOSOR947365-1G APOSOR947365-250MG
5-Chloro-2-formylbenzonitrile 95%
5-Chloro-2-formylbenzonitrile
Formel: C₈H₄ClNO
Molecular Weight: 165,576 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10699538
CAS-Nummer: 77532-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR