2-Methylbenzo[d]oxazol-5-ol 95%
Lieferant: Apollo Scientific
OR948086-1G
OR948086-250MG
APOSOR948086-1GEA
174
EUR
APOSOR948086-1G
APOSOR948086-250MG
2-Methylbenzo[d]oxazol-5-ol 95%
2-Methylbenzo[d]oxazol-5-ol
Formel:
C₈H₇NO₂ Molecular Weight: 149.147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11111705 CAS-Nummer: 23997-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...