5-Chloro-1-tetralone 98%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-3,4-dihydronaphthalen-1(2H)-one

OR948172-1G OR948172-5G
APOSOR948172-1GEA 53.8 EUR
APOSOR948172-1G APOSOR948172-5G
5-Chloro-1-tetralone 98%
5-Chloro-1-tetralone
Formel: C₁₀H₉ClO
Molecular Weight: 180,631 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08544416
CAS-Nummer: 26673-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR