(S)-2-Amino-2-(3-chlorophenyl)ethanol 97%

Lieferant: Apollo Scientific

OR948187-1G
APOSOR948187-1GEA 59.6 EUR
APOSOR948187-1G
(S)-2-Amino-2-(3-chlorophenyl)ethanol 97%
(S)-2-Amino-2-(3-chlorophenyl)ethanol
Formel: C₈H₁₀ClNO
Molecular Weight: 171,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09253677
CAS-Nummer: 663611-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR