1-(4-Bromophenyl)propan-1-amine 97%
Lieferant: Apollo Scientific
OR948235-1G
OR948235-250MG
APOSOR948235-1GEA
310
EUR
APOSOR948235-1G
APOSOR948235-250MG
1-(4-Bromophenyl)propan-1-amine 97%
1-(4-Bromophenyl)propan-1-amine
Formel:
C₉H₁₂BrN Molecular Weight: 214.102 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06655172 CAS-Nummer: 74877-09-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...