[1-(1-Methyl-1H-pyrazol-4-yl)ethyl]amine

Lieferant: Apollo Scientific
Gefahr

OR948344-1G OR948344-5G
APOSOR948344-1GEA 126 EUR
APOSOR948344-1G APOSOR948344-5G
[1-(1-Methyl-1H-pyrazol-4-yl)ethyl]amine
[1-(1-Methyl-1H-pyrazol-4-yl)ethyl]amine
Formel: C₆H₁₁N₃
Molecular Weight: 125,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864498
CAS-Nummer: 911788-33-5
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR