Boc-(R)-2-Cyclobutylglycine 97%

Lieferant: Apollo Scientific

OR948353-1G OR948353-250MG OR948353-500MG
APOSOR948353-1GEA 810 EUR
APOSOR948353-1G APOSOR948353-250MG APOSOR948353-500MG
Boc-(R)-2-Cyclobutylglycine 97%
Boc-(R)-2-Cyclobutylglycine
Formel: C₁₁H₁₉NO₄
Molecular Weight: 229,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659974
CAS-Nummer: 155905-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR