1-Methylcyclobutan-1-amine hydrochloride
Lieferant: Apollo Scientific
Synonyme:
1-Methylcyclobutanamine hydrochloride
OR948465-10G
OR948465-1G
OR948465-5G
APOSOR948465-10GEA
386
EUR
APOSOR948465-10G
APOSOR948465-1G
APOSOR948465-5G
1-Methylcyclobutan-1-amine hydrochloride
1-Methylcyclobutan-1-amine hydrochloride
Formel:
C₅H₁₁N·ClH Molecular Weight: 121,608 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09997048 CAS-Nummer: 174886-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...