5-Bromo-2-chloro-N4-ethylpyridine-3,4-diamine ≥98%

Lieferant: Apollo Scientific

OR948787-100MG OR948787-1G OR948787-250MG
APOSOR948787-100MGEA 526 EUR
APOSOR948787-100MG APOSOR948787-1G APOSOR948787-250MG
5-Bromo-2-chloro-N4-ethylpyridine-3,4-diamine ≥98%
5-Bromo-2-chloro-N4-ethylpyridine-3,4-diamine
Formel: C₇H₉BrClN₃
Molecular Weight: 250,523 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17015882
CAS-Nummer: 842144-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR