4-Chloro-1H-indole-7-carboxylic acid 97%

Lieferant: Apollo Scientific

OR949059-100MG OR949059-1G OR949059-250MG
APOSOR949059-100MGEA 306 EUR
APOSOR949059-100MG APOSOR949059-1G APOSOR949059-250MG
4-Chloro-1H-indole-7-carboxylic acid 97%
4-Chloro-1H-indole-7-carboxylic acid
Formel: C₉H₆ClNO₂
Molecular Weight: 195.602 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20657673
CAS-Nummer: 875305-77-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR