6-Bromo-3-methoxy-1H-indazole 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR949351-1G
OR949351-250MG
APOSOR949351-1GEA
219
EUR
APOSOR949351-1G
APOSOR949351-250MG
6-Bromo-3-methoxy-1H-indazole 98%
6-Bromo-3-methoxy-1H-indazole
Formel:
C₈H₇BrN₂O Molecular Weight: 227,058 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23106083 CAS-Nummer: 938062-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...