2-Propanoyloxybenzoic acid 95%
Lieferant: Apollo Scientific
OR949661-1G
OR949661-250MG
APOSOR949661-1GEA
0
EUR
APOSOR949661-1G
APOSOR949661-250MG
2-Propanoyloxybenzoic acid 95%
2-Propanoyloxybenzoic acid
Formel:
C₁₀H₁₀O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01684366 CAS-Nummer: 6328-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...