6-Methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine hydrochloride 95%

Lieferant: Apollo Scientific

OR949833-1G OR949833-250MG
APOSOR949833-1GEA 719 EUR
APOSOR949833-1G APOSOR949833-250MG
6-Methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine hydrochloride 95%
6-Methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine hydrochloride
Formel: C₉H₁₁NO·ClH
Molecular Weight: 185,651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09026861
CAS-Nummer: 1095270-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR