Fmoc-N-methyl-D-norleucine 97%

Lieferant: Apollo Scientific

Synonyme: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)hexanoic acid

OR950165-1G OR950165-500MG OR950165-5G
APOSOR950165-1GEA 281 EUR
APOSOR950165-1G APOSOR950165-500MG APOSOR950165-5G
Fmoc-N-methyl-D-norleucine 97%
Fmoc-N-methyl-D-norleucine
Formel: C₂₂H₂₅NO₄
Molecular Weight: 367,438 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366865
CAS-Nummer: 1217482-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR