1-(4-Methoxyphenyl)-2-methylpropan-1-ol 95%
Lieferant: Apollo Scientific
OR950276-1G
OR950276-250MG
APOSOR950276-1GEA
227
EUR
APOSOR950276-1G
APOSOR950276-250MG
1-(4-Methoxyphenyl)-2-methylpropan-1-ol 95%
1-(4-Methoxyphenyl)-2-methylpropan-1-ol
Formel:
C₁₁H₁₆O₂ Molecular Weight: 180,244 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11934907 CAS-Nummer: 18228-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...