2-(2-Methoxypyridin-4-yl)acetic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR950553-250MG
OR950553-1G
APOSOR950553-250MGEA
162
EUR
APOSOR950553-250MG
APOSOR950553-1G
2-(2-Methoxypyridin-4-yl)acetic acid 95%
2-(2-Methoxypyridin-4-yl)acetic acid
Formel:
C₈H₉NO₃ Molecular Weight: 167,162 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10698166 CAS-Nummer: 464152-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...