3-Bromo-6-methyl-5,6-dihydro-7H-pyrrolo[3,4-b]pyridin-7-one 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-6-methyl-5H-pyrrolo[3,4-b]pyridin-7(6H)-one
OR950728-1G
OR950728-250MG
APOSOR950728-1GEA
799
EUR
APOSOR950728-1G
APOSOR950728-250MG
3-Bromo-6-methyl-5,6-dihydro-7H-pyrrolo[3,4-b]pyridin-7-one 95%
3-Bromo-6-methyl-5,6-dihydro-7H-pyrrolo[3,4-b]pyridin-7-one
Formel:
C₈H₇BrN₂O Molecular Weight: 227,058 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20229834 CAS-Nummer: 1254319-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...