N-[2-(Hydroxymethyl)phenyl]acetamide 95%
Lieferant: Apollo Scientific
Synonyme:
N-(2-(Hydroxymethyl)phenyl)acetamide
OR950936-1G
OR950936-5G
APOSOR950936-1GEA
78.3
EUR
APOSOR950936-1G
APOSOR950936-5G
N-[2-(Hydroxymethyl)phenyl]acetamide 95%
N-[2-(Hydroxymethyl)phenyl]acetamide
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,189 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14648693 CAS-Nummer: 20939-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...