4-Chlorobenzoylformic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Chlorophenyl)-2-oxoacetic acid

OR951002-1G OR951002-250MG
APOSOR951002-1GEA 502 EUR
APOSOR951002-1G APOSOR951002-250MG
4-Chlorobenzoylformic acid 95%
4-Chlorobenzoylformic acid
Formel: C₈H₅ClO₃
Molecular Weight: 184,577 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02656397
CAS-Nummer: 7099-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR