4-Cyano-2-ethylpyridine 95%
Lieferant: Apollo Scientific
Synonyme:
2-Ethylisonicotinonitrile
OR951014-1G
OR951014-250MG
APOSOR951014-1GEA
415
EUR
APOSOR951014-1G
APOSOR951014-250MG
4-Cyano-2-ethylpyridine 95%
4-Cyano-2-ethylpyridine
Formel:
C₈H₈N₂ Molecular Weight: 132.163 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12546403 CAS-Nummer: 1531-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...