5-Amino-3-methyl-isoxazole-4-carboxylic acid ethyl ester 96%

Lieferant: Apollo Scientific

Synonyme: Ethyl 5-amino-3-methylisoxazole-4-carboxylate

OR951167-1G OR951167-250MG
APOSOR951167-1GEA 180 EUR
APOSOR951167-1G APOSOR951167-250MG
5-Amino-3-methyl-isoxazole-4-carboxylic acid ethyl ester 96%
5-Amino-3-methyl-isoxazole-4-carboxylic acid ethyl ester
Formel: C₇H₁₀N₂O₃
Molecular Weight: 170,166 g/mol
Schmelzpunkt: 133…135 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD03820571
CAS-Nummer: 25786-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR